Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-amino-3-(pyridin-4-yl)propanoic acid dihydrochloride | 178933-04-5 | MFCD13184932 | 98.0% | 239.10 g/mol | C8H12Cl2N2O2 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0224051G

Encompass_Preferred

(S)-2-Amino-3-(pyridin-4-yl)propanoic acid dihydrochloride is the S-enantiomer of a pyridinyl-substituted alanine derivative supplied as the dihydrochloride salt for research use. It is provided as a high-purity solid suitable for biochemical, pharmacological, and medicinal chemistry studies.

  • Stereochemically pure (S)-enantiomer.
  • Supplied as the dihydrochloride salt to aid solubility.
  • High purity: 98.0%.
  • Molecular weight: 239.10 g/mol.
  • Chemical formula: C8H12Cl2N2O2.
  • Available in multiple pack sizes, including 1 g.

Catalog No. 50-002-99602


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.