Learn More
Medchemexpress LLC (S)-2-amino-3-(pyridin-4-yl)propanoic acid dihydrochloride | 178933-04-5 | MFCD13184932 | 98.4% | 239.10 g/mol | C8H12Cl2N2O2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW022405
Encompass_Preferred
(S)-2-Amino-3-(pyridin-4-yl)propanoic acid dihydrochloride is the dihydrochloride salt of an S-configured pyridinylalanine derivative provided for research use. It is supplied as a characterized crystalline solid with analytical documentation supporting identity and purity, and is used as a synthetic building block and reagent in biochemical and peptide-related studies.
  • High stereochemical purity suitable for chiral studies.
  • Validated purity by RP-HPLC for analytical confidence.
  • Supplied with COA, HNMR, and RP-HPLC documentation.
  • Available in multiple small-scale pack sizes for research workflows.
  • Stable crystalline solid convenient for storage and handling.
Catalog No. 50-003-03653