Learn More
Medchemexpress LLC (S)-2-amino-5-(tert-butoxy)-5-oxopentanoic acid | 2419-56-9 | MFCD00038580 | 98.0% | 203.24 g/mol | C9H17NO4 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW014742500G
Encompass_Preferred
(S)-2-Amino-5-(tert-butoxy)-5-oxopentanoic acid is a chiral glutamic acid derivative supplied as a solid for research applications. It serves as an amino acid building block for biochemical synthesis and pharmacological studies, and is characterized by its molecular composition and documented stability.
  • Chiral glutamic acid derivative for research use.
  • Useful as an amino acid building block in synthesis.
  • Molecular formula C9H17NO4 and molecular weight 203.24 g/mol.
  • Stated purity 98.0%.
  • Available in bulk solid quantities suitable for preparative work.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-003-02966