Learn More
Medchemexpress LLC (S)-2-(((benzyloxy)carbonyl)amino)pent-4-enoic acid | 78553-51-2 | MFCD02094548 | >98.0% | 249.27 g/mol | C13H15NO4 | 5 G

Supplier: Medchemexpress LLC HYW0404165G
(S)-2-(((benzyloxy)carbonyl)amino)pent-4-enoic acid is a Cbz-protected amino acid derivative used as a research building block and synthetic intermediate. It is provided as a solid for laboratory use, with documented identifiers and supporting safety documentation.
- has CAS number 78553-51-2
- molecular formula C13H15NO4
- molecular weight approx. 249.27 g/mol
- available in a 5 g packaged quantity
- used as an amino acid derivative and peptide synthesis intermediate
- safety data sheet and documentation available from the supplier
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.