Learn More
STA PHARMACEUTICAL US LLC (S)-2-(Fmoc-Amino)-4-Iodobutanoic Acid | 1 g | CAS 1932598-55-4 | InChIKey WUMHLUDKFHNQNJ-KRWDZBQOSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAA117881GM
(S)-2-(Fmoc-Amino)-4-Iodobutanoic Acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1932598-55-4
- MDL: No data
- InChIKey: WUMHLUDKFHNQNJ-KRWDZBQOSA-N
- Molecular Weight: 451.26
- Molecular Formula: C19H18INO4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-iodobutanoic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=CC=CC=C31)N[C@H](C(O)=O)CCI
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.