Learn More
Cayman Chemical S-2 MethndamIde 10mg
SDP
Supplier:  Cayman Chemical 9007610 MG
Encompass_Preferred
The second most potent CB1 receptor agonist in the methanandamide series (Ki = 26 nM for the CB1 receptor); less prone to FAAH inactivation and inhibits the murine vas deferens twitch response (IC50 = 47 nM)
Catalog No. 50-289-5811