Learn More
Medchemexpress LLC (S)-2-(Methoxycarbonylamino)-3,3-dimethylbutanoic acid | 162537-11-3 | 98.0% | C8H15NO4 | 100 G
SDP
Supplier:  Medchemexpress LLC HY59135100G
Encompass_Preferred
(S)-2-(Methoxycarbonylamino)-3,3-dimethylbutanoic acid is a leucine derivative widely used as an ergogenic supplement. It influences anabolic hormones, fuels exercise, and enhances mental performance during stress, while also preventing exercise-induced muscle damage.
  • Leucine derivative
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Enhances mental performance during stress
  • Prevents exercise induced muscle damage
  • Recognized as a beneficial ergogenic dietary substance
Catalog No. 50-004-34126