Learn More
Medchemexpress LLC (S)-2-(Methylamino)-3-phenylpropanoic acid | 2566-30-5 | 179.22 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW005388
Encompass_Preferred
The (S)-2-(Methylamino)-3-phenylpropanoic acid is a phenylalanine derivative. Amino acids and their derivatives are recognized as beneficial ergogenic dietary substances, commercially used as supplements influencing anabolic hormones, fuel supply during exercise, mental performance during stress-related tasks, and preventing exercise-induced muscle damage.
  • Phenylalanine derivative
  • Used as ergogenic supplements
  • Influences secretion of anabolic hormones
  • Aids fuel supply during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • Appears as a white to off-white solid
  • Soluble in water (40 mg/mL)
  • Supporting documentation is available
  • For research use only
Catalog No. 50-004-48201