Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-2-Methylpyrrolidine-2-carboxylic acid | 42856-71-3 | 98.0% | C6H11NO2 | 10 G
SDP

Supplier:  Medchemexpress LLC HY7696210G

Encompass_Preferred

(S)-2-Methylpyrrolidine-2-carboxylic acid is a proline derivative recognized as a beneficial ergogenic dietary substance. It influences anabolic hormone secretion, the supply of fuel during exercise, and mental performance during stress-related tasks, while also helping prevent exercise-induced muscle damage.

  • Proline derivative
  • Recognized as an ergogenic dietary substance
  • Influences anabolic hormone secretion
  • Supports fuel supply during exercise
  • Enhances mental performance during stress-related tasks
  • Aids in preventing exercise-induced muscle damage
  • Solid appearance, white to off-white
  • Soluble in water at 100 mg/mL
  • Recommended storage at 4°C, protected from light and under nitrogen

Catalog No. 50-004-33996


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.