Learn More
Medchemexpress LLC (S)-(+)-2-phenylglycine methyl ester hydrochloride | 15028-39-4 | MFCD00077158 | 99.9% | 201.65 g/mol | C9H12ClNO2 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW019714500G
Encompass_Preferred
(S)-(+)-2-Phenylglycine methyl ester hydrochloride is the S-enantiomer of 2-phenylglycine methyl ester supplied as its hydrochloride salt. It is a white to off-white solid used as a chiral building block and intermediate in medicinal chemistry and asymmetric synthesis, provided with technical and safety documentation for research use.
  • Is the S-enantiomer of 2-phenylglycine methyl ester.
  • Presented as the hydrochloride salt for improved handling and stability.
  • White to off-white solid suitable as a chiral building block.
  • High purity, typically 99.89%.
  • Molecular weight 201.65 g/mol; formula C9H12ClNO2.
  • Available in gram-scale quantities for synthesis and scale-up.
Catalog No. 50-003-00098