Learn More
Medchemexpress LLC (S)-(+)-2-Phenylglycinol | 20989-17-7 | 99.29% | 137.18 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY41414
Encompass_Preferred
(S)-(+)-2-Phenylglycinol is a biochemical reagent suitable for use as a biological material or organic compound in life science research.
  • High purity: 99.29%
  • Available in multiple quantities (25g to 1kg)
  • Good solubility in DMSO (100 mg/mL)
  • Stable under various storage conditions
  • Applicable for life science research
  • Detailed dissolution protocols for in vitro and in vivo studies
Catalog No. 50-004-03032