Learn More
Medchemexpress LLC S-22153 25mg | 180304-07-8 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11496225MG
Encompass_Preferred
S-22153 is a potent melatonin receptor antagonist with EC50 values of 19 nM 4 6 nM for hMT1 and hMT2 melatonin receptor respectively S-22153 has Ki values of 8 6 nM (CHO cells) and 16 3 nM (HEK cells) for hMT1 and 6 0 nM (CHO cells) and 8 2 nM (HEK cells) for hMT2 S-22153 is a specific ligand of MT1 and MT2 melatonin receptors subtypes[1][2]
Catalog No. 50-002-28506