Learn More
Medchemexpress LLC S-22153 50mg | 180304-07-8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11496250MG
Encompass_Preferred
S-22153 is a melatonin MT1/MT2 receptor antagonist research compound (CAS 180304-07-8) supplied as a high-purity powder for laboratory pharmacology and receptor-binding studies.
  • High purity suitable for in vitro assays.
  • Available in small pack sizes for screening and dose-ranging.
  • Stable when stored under recommended conditions.
  • Molecular formula C14H17NOS; molecular weight 247.36 g/mol.
Catalog No. 50-003-07561