Learn More
Cayman Chemical S-2238hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3494410 MG
Encompass_Preferred
A chromogenic substrate for thrombin; enzymatic cleavage releases pNA which can be quantified by colorimetric detection at 405 nm as a measure of thrombin activity
Catalog No. 50-291-2297