Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-3,4-dicarboxyphenylglycine | 201730-11-2 | MFCD03453633 | ≥98.0% | 239.18 g/mol | C10H9NO6 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1075165MG

Encompass_Preferred

(S)-3,4-dicarboxyphenylglycine is a research-grade small molecule that functions as a selective agonist of metabotropic glutamate receptor 8a (mGluR8a). It is used in receptor pharmacology and preclinical neuroscience studies and has been characterized in cell-based assays for potency and selectivity.

  • Selective agonist of mGluR8a with EC50 31 nM in AV12-664 cells.
  • High purity suitable for research use (≥98.0%).
  • Molecular weight 239.18 g/mol.
  • CAS number 201730-11-2 for unambiguous identification.
  • Off-white to light yellow solid physical form.
  • Powder stable with recommended cold storage for long-term retention.
  • Appropriate for receptor pharmacology, signaling, and preclinical studies.

Catalog No. 50-002-39990


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.