Learn More
Medchemexpress LLC (S)-3,4-Dihydroxybutyric acid (lithium hydrate) | 98.1% | 144.05 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY113304A10MG
Encompass_Preferred
(S)-3,4-Dihydroxybutyric acid (lithium hydrate) is an endogenous metabolite found in human urine. It is excreted in higher concentrations in patients who have a deficiency in succinic semialdehyde dehydrogenase (SSADH).
  • Endogenous metabolite
  • For research use only (not for sale to patients)
Catalog No. 50-004-35624