Learn More
Medchemexpress LLC (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(allyloxy)-4-oxobutanoic acid | 144120-53-6 | MFCD00467715 | 98.7% | 395.41 | C22H21NO6 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0170695G
Encompass_Preferred
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(allyloxy)-4-oxobutanoic acid is an Fmoc-protected L-aspartic acid derivative used as a synthetic intermediate in peptide and organic synthesis. It provides an N-Fmoc protecting group and an allyl-protected side chain for orthogonal deprotection strategies.
  • Protected aspartic acid derivative suitable for peptide synthesis.
  • Fmoc protection on the amino group compatible with standard Fmoc SPPS.
  • Allyl-protected side chain enables orthogonal deprotection workflows.
  • White to off-white solid, convenient handling and storage.
  • High purity (98.7%) appropriate for research use.
  • Molecular formula C22H21NO6; molecular weight 395.41 g/mol.
Catalog No. 50-003-00353