Learn More
Medchemexpress LLC (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde | 102308-32-7 | 98.6% | C11H19NO4 | 50 G
SDP
Supplier:  Medchemexpress LLC HY6934850G
Encompass_Preferred
(S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde is a chiral compound used as an intermediate in asymmetric synthesis and organic synthesis. As a polar aldehyde substrate, it enhances polarity, solubility, and hydrogen bond formation.
  • Chiral compound for synthesis
  • Intermediate in asymmetric synthesis
  • Applied in organic synthesis
  • Enhances compound polarity
  • Improves compound solubility
  • Facilitates hydrogen bond formation
Catalog No. 50-004-34761