Learn More
Medchemexpress LLC (S)-3-(Boc-amino)-4-phenylbutyric acid | 51871-62-6 | MFCD01076271 | >=95.0% | 279.33 g/mol | C15H21NO4 | 10 G

Supplier: Medchemexpress LLC HYW04247810G
(S)-3-(Boc-amino)-4-phenylbutyric acid is a Boc-protected phenylalanine derivative supplied as a solid powder for research use. It is commonly used as a protected amino-acid intermediate in peptide synthesis and small-molecule research.
- Solid powder physical state
- Purity ≥95.0%
- Molecular weight 279.33 g/mol
- Chemical formula C15H21NO4
- Available in multiple pack sizes, including 10 g
- Storage: powder -20°C for 3 years, 4°C for 2 years
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.