Learn More
Medchemexpress LLC (S)-3-cyclopropyl-4,7,7-trimethyl-4-phenyl-2,4,6,7,8,9-hexahydro-pyrazolo[3,4-b]quinolin-5-one | 1597439-87-6 | 99.6% | 347.45 g/mol | C22H25N3O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117822100MG
Encompass_Preferred
BRD0209 is a potent, reversible ATP-competitive inhibitor of glycogen synthase kinase 3 alpha and beta (GSK3α/β) with low-nanomolar potency. Provided as a high-purity solid for research use, it is suitable for in vitro biochemical assays, cell-based studies, and compound screening.
  • Potent GSK3α/β inhibition (GSK3α IC50 = 19 nM; GSK3β IC50 = 5 nM).
  • High purity for reliable assay performance.
  • Solid form with defined storage recommendations for stability.
  • Supplied as a 100 mg quantity for laboratory use.
  • Suitable for biochemical and cell-based research applications.
Catalog No. 50-002-27255