Learn More
Medchemexpress LLC S -3-N-Cbz-Amino-su 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW097106
Encompass_Preferred
(S)-3-N-Cbz-Amino-succinimide (Compound 1d) is an antiepileptic agent that can inhibit pentylenetetrazole (PTZ) and maximal electroshock (MES)-induced tonic convulsions in mice[1]
Catalog No. 50-002-04651