Learn More
Medchemexpress LLC HY-106992 1mg Medchemexpress, S 3304 CAS:203640-27-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1069921MG
Encompass_Preferred
Medchemexpress, HY-106992 1mg S 3304 CAS:203640-27-1 S 3304 is a novel matrix metalloproteinases (MMP) inhibitor specific for MMP-2 and MMP-9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8361