Learn More
Strem, An Ascensus Company CAS# 325168-88-5. 100mg. (S)-(+)-4,12-Bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane, min. 97% CTH-(S)-3,5-xylyl-PHANEPHOS. MFCD03840578
SDP
Supplier:  Strem, An Ascensus Company 150731100MG
Encompass
CAS# 325168-88-5. 100mg. (S)-(+)-4,12-Bis(di(3,5-xylyl)phosphino)-[2.2]-paracyclophane, min. 97% CTH-(S)-3,5-xylyl-PHANEPHOS. MFCD03840578. Molecular Weight: 688.86. Molecular Formula: C48H50P2. Color/form: white pwdr. Strem# 15-0731. http://www.strem.com/catalog/v/15-0731/
Catalog No. 50-901-09967