Learn More
Medchemexpress LLC (S)-4-(8-Amino-3-(1-(but-2-ynoyl)pyrrolidin-2-yl)imidazo[1,5-a]pyrazin-1-yl)-N-(pyridin-2-yl-d4)benzamide | 2699608-18-7 | >=98.0% | 469.53 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17600S1MG
Encompass_Preferred
Acalabrutinib-d4 (ACP-196-d4) is a deuterium labeled Acalabrutinib, an orally active, irreversible, and highly selective second-generation BTK inhibitor. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used as a tracer for quantitation during drug development.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes to affect pharmacokinetic and metabolic profiles.
  • Suitable for research use only.
Catalog No. 50-003-87530