Learn More
Medchemexpress LLC (S)-4-Benzyl-3-(5-bromopentanoyl)oxazolidin-2-one | 156699-37-5 | 99.9% | 340.22 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY42520
Encompass_Preferred
(S)-4-Benzyl-3-(5-bromopentanoyl)oxazolidin-2-one is a drug intermediate for the synthesis of various active compounds. It is stable under recommended storage conditions.
  • Harmful if swallowed.
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.
  • Avoid breathing dust, fume, gas, mist, vapours, or spray.
  • Wash hands thoroughly after handling.
  • Do not eat, drink, or smoke when using this product.
  • Use only outdoors or in a well-ventilated area.
  • Wear protective gloves, protective clothing, eye protection, and face protection.
  • Keep container tightly sealed in a cool, well-ventilated area.
  • Keep away from direct sunlight and sources of ignition.
  • Incompatible with strong acids, alkalis, and strong oxidising or reducing agents.
Catalog No. 50-004-02290