Learn More
Medchemexpress LLC S-(4-Hydroxybenzyl)glutathione | 129636-38-0 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN811510MG
Encompass_Preferred
S-(4-Hydroxybenzyl)glutathione is a glutathione derivative. It has been shown to inhibit the in vitro binding of kainic acid to brain glutamate receptors, with an IC50 of 2 μM.
  • It is a glutathione derivative.
  • It inhibits the in vitro binding of kainic acid to brain glutamate receptors.
  • It has an IC50 of 2 μM for inhibiting kainic acid binding.
Catalog No. 50-004-17613