Learn More
Cayman Chemical S-4-nItro-BlebbIstatIn 5mg
SDP
Supplier:  Cayman Chemical 241715 MG
Encompass_Preferred
A more stable and less phototoxic form of (–)-blebbistatin; less inherent fluorescence, more photostable, and less phototoxic than (–)-blebbistatin while retaining the in vitro and in vivo activity; has the same stereochemistry as (–)-blebbistatin
Catalog No. 50-293-6534