Learn More
Medchemexpress LLC (S)-4-phenyloxazolidin-2-one | 99395-88-7 | MFCD00043396 | 100.0% | 163.17 g/mol | C9H9NO2 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW019727500G
Encompass_Preferred
(S)-4-Phenyloxazolidin-2-one is a chiral 2-oxazolidinone reagent used as an intermediate and building block in stereoselective organic synthesis and medicinal chemistry research. The material is supplied as a high-purity biochemical reagent (CAS 99395-88-7) intended for research use.
  • High purity suitable for research applications.
  • Molecular formula C9H9NO2 and molecular weight 163.17 g/mol.
  • Useful as a chiral building block for asymmetric synthesis and method development.
  • Offered in multiple pack sizes, including a 500 g option for scale-up.
  • Provided for laboratory research and development purposes.
Catalog No. 50-003-00100