Learn More
Strem, An Ascensus Company CAS# 210169-57-6. 1g. (S)-(-)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole, min. 98% (S)-(-)-DM-SEGPHOS. MFCD09753008
SDP
Supplier:  Strem, An Ascensus Company 1504791G
Encompass
CAS# 210169-57-6. 1g. (S)-(-)-5,5'-Bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole, min. 98% (S)-(-)-DM-SEGPHOS. MFCD09753008. Molecular Weight: 722.79. Molecular Formula: C46H44O4P2. Color/form: off-white to pale yellow pwdr. Strem# 15-0479. http://www.strem.com/catalog/v/15-0479/
Catalog No. 50-138-5373