Learn More
Medchemexpress LLC (S)-5-fluoro-6-chloro-alpha-methylisotryptamine | 169675-08-5 | 99.5% | 226.68 g/mol | C11H12ClFN2 | 10MG

Supplier: Medchemexpress LLC HY12383810MG
Ro60-0175 is a small-molecule research compound and a potent, selective agonist of the serotonin 5-HT2C receptor, used in preclinical pharmacology and behavioral studies (reported to reduce self-administration). The product is supplied as the free-base solid for in vitro and in vivo research.
- Potent, selective 5-HT2C receptor agonist.
- High purity (99.52%).
- Solid form; white to light yellow appearance.
- Molecular weight 226.68 g/mol; formula C11H12ClFN2.
- CAS 169675-08-5 for unambiguous identification.
- Recommended storage: 4°C for solid; protect from light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.