Learn More
Strem, An Ascensus Company CAS# 503538-68-9. 500mg. S-(-)-6,6'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin, min. 97% (S)-SYNPHOS™. MFCD05861608
SDP
Supplier:  Strem, An Ascensus Company 150491500MG
Encompass
CAS# 503538-68-9. 500mg. S-(-)-6,6'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin, min. 97% (S)-SYNPHOS™. MFCD05861608. Molecular Weight: 638.63. Molecular Formula: C40H32O4P2. Color/form: white pwdr. Strem# 15-0491. http://www.strem.com/catalog/v/15-0491/
Catalog No. 50-901-09934