Learn More
Medchemexpress LLC (S)-ACE-OH | 99.2% | 328.47 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1693255MG
Encompass_Preferred
(S)-ACE-OH is a molecular glue with anticancer activity. It promotes nucleoporin degradation and disruption of nucleocytoplasmic trafficking by inducing the interaction between E3 ubiquitin ligase TRIM21 and nucleoporin NUP98.
  • Anticancer activity
  • Molecular glue
  • Promotes nucleoporin degradation
  • Disrupts nucleocytoplasmic trafficking
  • Induces interaction between E3 ubiquitin ligase TRIM21 and nucleoporin NUP98
  • Targeted therapy for PROTAC molecular glues
Catalog No. 50-003-90963