Learn More
Medchemexpress LLC S-acetyl-PEG2-Boc | 1820641-93-7 | C13H24O5S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY141345500MG
Encompass_Preferred
S-acetyl-PEG2-Boc is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs are compounds consisting of two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. PROTACs operate by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55716