Learn More
Cayman Chemical S-AdenosylhomocysteIn 10mg
SDP
Supplier:  Cayman Chemical 1360310 MG
Encompass_Preferred
An amino acid derivative and an intermediate, by-product, or modulator of several metabolic pathways, including the activated methyl cycle and cysteine biosynthesis; also a product of SAM-dependent methylation of biological molecules, including DNA, RNA, and histones, and other proteins
Catalog No. 50-295-5408