Learn More
Cayman Chemical S-AdenosylhomocysteIn-d4 500UG
SDP
Supplier:  Cayman Chemical 9000372500 UG
Encompass_Preferred
An internal standard for the quantification of SAH by GC- or LC-MS
Catalog No. 50-289-2631