Learn More
Medchemexpress LLC S-Allyl-L-cysteine | 21593-77-1 | 98.6% | 161.22 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01357310MM/1ML
Encompass_Preferred
S-Allyl-L-cysteine is an organosulfur compound found in aged garlic extract (AGE). It exhibits diverse biological activities, including neurotrophic, anti-cancer, and anti-inflammatory effects. Research indicates its potential in protecting against cell death and regulating cellular processes.
  • Protects against amyloid-protein and tunicamycin-induced cell death.
  • Suppresses reactive oxygen species (ROS) levels.
  • Regulates apoptotic signals.
  • Demonstrates neurotrophic activity.
  • Exhibits anti-cancer activity.
  • Possesses anti-inflammatory activity.
Catalog No. 50-004-54458