Learn More
Medchemexpress LLC S-Allyl-L-cysteine | 21593-77-1 | 98.6% | 161.22 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0135735MG
Encompass_Preferred
S-Allyl-L-cysteine is an organosulfur compound found in Aged Garlic Extract (AGE). It exhibits various biological effects, including neurotrophic, anti-cancer, and anti-inflammatory activities. This product is intended for research use only.
  • Protects against amyloid-protein and tunicamycin-induced cell death in differentiated PC12 cells.
  • Regulates apoptotic signals by suppressing the transcription and protein expression of caspase-3.
  • Decreases Aβ-induced levels of reactive oxygen species (ROS).
Catalog No. 50-004-54525