Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific (S)-(+)-alpha-Methoxy-alpha-trifluoromethyl-phenylacetyl chloride, 20445-33-4, MFCD00067105, 1g

Supplier:  Matrix Scientific 0071701G

Encompass_Preferred

Molecular Formula C10H8ClF3O2, Purity 99%, Molecular Weight 252.62, Boiling Point 213-214

Catalog No. 50-012-0197


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.