Learn More
Medchemexpress LLC (S)-AM-9022 | 2446872-47-3 | 99.9% | 578.67 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY162110A1MG
Encompass_Preferred
(S)-AM-9022 is the S-enantiomer of AM-9022. It is an orally active, potent, and selective KIF18A inhibitor, making it suitable for cancer research.
  • S-enantiomer of AM-9022
  • Orally active
  • Potent and selective KIF18A inhibitor
  • Suitable for cancer research
Catalog No. 50-003-92282