Learn More
Medchemexpress LLC (βS)-β-amino-4-[(3,4-dichlorophenyl)methoxy]-β-methyl-benzenebutanol | 2748196-63-4 | 97.0% | 354.27 g/mol | C18H21Cl2NO2 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12601525MG
Encompass_Preferred
P053 is a potent, selective, non-competitive inhibitor of ceramide synthase 1 (CerS1) (IC50 ≈ 0.5 μM), intended for use as a research reagent to study CerS1 function, mitochondrial fatty-acid oxidation in muscle, and adiposity regulation. For research use only.
  • Potent CerS1 inhibition with IC50 ≈ 0.5 μM.
  • Selective for CerS1 over other ceramide synthase isoforms.
  • High purity (97.0%) suitable for biochemical assays.
  • Available in small milligram pack sizes for research flexibility.
  • Suitable for biochemical and cellular studies of lipid metabolism.
Catalog No. 50-002-36451