Learn More
Abcam (S)-(-)-Atenolol, beta1 adrenoceptor antagonist, 10MG
Supplier:  Abcam AB12085610MG
Encompass_Preferred
MW 266.34 g/mol, Purity >98%. Selective β₁ adrenoceptor antagonist. Orally active. Limited ability to cross the blood-brain barrier. Antihypertensive activity in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7194