Learn More
Medchemexpress LLC (S)-Benzyl 27-benzyl-1-(9H-fluoren-9-yl) hexaoxahexaazahexatriacontan-oate | 2914227-56-6 | 99.7% | 983.07 | C51H62N6O14 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1568105MG
Encompass_Preferred
Fmoc-PEG4-GGFG-CH2-O-CH2-Cbz is an Fmoc-protected, PEG4-containing cleavable linker used for the synthesis of antibody-drug conjugates (ADCs). It is supplied as a high-purity solid reagent for research-scale bioconjugation and linker-payload assembly.
  • Fmoc-protected functionality for standard peptide coupling workflows.
  • Four-unit PEG spacer to improve solubility and provide distance between antibody and payload.
  • Cleavable linker chemistry suitable for assembly and payload release strategies.
  • High purity (>99.6%) solid, suitable for research and development applications.
  • Recommended storage: 4°C sealed; in solvent store at -80°C for long-term storage.
Catalog No. 50-002-04370