Learn More
Strem, An Ascensus Company CAS# 76189-56-5. 250MG. (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% (S)-(-)-BINAP. MFCD00010805
SDP
Supplier:  Strem, An Ascensus Company 150151250MG
Encompass
CAS# 76189-56-5. 250MG. (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% (S)-(-)-BINAP. MFCD00010805. Molecular Formula: C44H32P2. Molecular Weight: 622.70. Color/Form: white to off-white pwdr. Strem# 15-0151. www.strem.com/catalog/v3/15-0151/
Catalog No. 50-196-3575