Learn More
Bellco Glass, Inc. Screw Cap 45mm ErlenmeyerFlask, 250mL Graduated
SDP
Supplier:  Bellco Glass, Inc. 251500250
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for all cell culture and media storage purposes. Provides flexible secure seal without a liner. Supplied with gl45 liner-less screw cap and pouring rings. Graduated to approximate volumes.. MADE IN THE USA Size: 250mL Dimensions: 80x117mm
Catalog No. 50-194-5483