Learn More
Bellco Glass, Inc. Screw Cap Leighton Tube,16x93mm
SDP
Supplier:  Bellco Glass, Inc. 190416095
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes long skirted screw cap with nontoxic white rubber liner. Extra long working area yet short overall length for convenient handling. The short neck provides easy entry and removal of cover slips. MADE IN THE USA
Catalog No. 50-194-4675