Learn More
Bellco Glass, Inc. S/Cap Cult Tube Complete, 16x150mm Bulk-Pack - No logo
SDP
Supplier:  Bellco Glass, Inc. 201216000
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA Screw cap included Size: 16 x 150mm
Catalog No. 50-194-5296