Learn More
Bellco Glass, Inc. S/Cap Grad Erlenmeyer Flask ,4000mL With 38mm Cap
SDP
Supplier:  Bellco Glass, Inc. 251404000
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes. . MADE IN THE USA Size: 4L Cap: 38-415 Dimensions: 205x350mm
Catalog No. 50-194-5481