Learn More
Cygnus Technologies S. cerevisiae HCP ELISA Kit
Supplier:  Cygnus Technologies F135
Encompass
This kit is for determining the presence of host cell protein impurities in products recombinantly expressed in S. cerevisiae. Our assay qualification has determined that this kit has an LOD of ~0.5 ng/ml.
Catalog No. NC1431077