Learn More
Medchemexpress LLC (S)-(-)-Citronellal | 5949-05-3 | 99.7% | 154.25 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY111664A500MG
Encompass_Preferred
(S)-(-)-Citronellal is a monoterpenoid compound derived from a biosynthetic pathway where mevalonic acid serves as the sole carbon source. It is found in essential oils of *Corymbia citriodora* and *Cymbopogon nardus*. (S)-(-)-Citronellal can also function as a starting material in the cyclization process.
  • Monoterpenoid compound.
  • Derived from a biosynthetic pathway with mevalonic acid.
  • Present in essential oils of *Corymbia citriodora* and *Cymbopogon nardus*.
  • Can be used as a starting material for cyclization.
  • For research use only.
Catalog No. 50-004-39256