Learn More
Medchemexpress LLC (S)-crizotinib | 1374356-45-2 | MFCD22126092 | 99.8% | 450.34 | C21H22Cl2FN5O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10054910MM 1ML
Encompass_Preferred
(S)-Crizotinib is the S-enantiomer of crizotinib supplied for research use as a stereochemically defined small molecule active against MTH1 and used in studies of DNA/RNA synthesis and apoptosis. It is provided as a ready-to-use solution in DMSO or as solid material for biochemical and cellular assays.
  • Stereochemically pure S-enantiomer suitable for enantiomer-specific studies.
  • Selective MTH1 inhibitor for biochemical and cellular assays.
  • High purity, 99.8% (HPLC), suitable for research applications.
  • Available as a 10 mM solution in DMSO (1 ML) and in multiple solid sizes.
  • Molecular weight 450.34 and formula C21H22Cl2FN5O for reference.
Catalog No. 50-002-11820